Trifluoroacetic acid 0.05% (v/v) in aqueous solution, LiChrosolv® for HPLC, Supelco®
Catalog # 4.80170.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:114.02 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-929-3
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169