

Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv® for liquid chromatography, Supelco®
Catalog # 4.80448.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (trifluoroacetic acid):0.095 - 0.105 % (v/v)
Specification Test Results
About this item
:
Density:0.786 g/cm3 (20 °C)
Flash Pt:2 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:200-929-3
:GHS02
:GHS07
:75-05-8