Order Entry
Czech Republic
ContactUsLinkComponent
Semicarbazide hydrochloride, Sigma-Aldrich®
Toxic
Semicarbazide hydrochloride, Sigma-Aldrich®
Catalog # SIAL84940-100G
Supplier:  Merck
CAS Number:  
Toxic
Semicarbazide hydrochloride, Sigma-Aldrich®
Catalog # SIAL84940-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :111.53 g/mol
Merck Index:12,08588
Melting Pt:174…178 °C
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:209-247-0
UN:2811
 :GHS06
CAS Number:563-41-7
Formula:CH₆ClN₃O
MDL Number:MFCD00013009