

Semicarbazide hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:563-41-7
:
Formula:CH₆ClN₃O
MDL Number:MFCD00013009
:111.53 g/mol
Merck Index:14,8444
Melting Pt:∼ 176 °C (dec.)
Storage Temperature:Ambient
EINECS:209-247-0
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together