Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Semicarbazide hydrochloride 99+%
 
undefined
 
 
Semicarbazide hydrochloride 99+%

 

  • Appearance:
    White Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    170 to 178 °C
  • Sulfated ash:
    ≤0.1 %
  • Solubility:
    (10 % in water) Clear colorless
  • Titration with KIO3:
    ≥99.0 %

 

 

Frequently Bought Together