
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1465-25-4
:
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
:259.18 g/mol
Merck Index:12,06496
Melting Pt:188…190 °C
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (calculated on dried substance):Min. 98.0 %
- IR Spectrum:Passes test
- Sensitivity to sulphanilamide:Passes test
- Solubility:Passes test
- Loss on drying (80°C; 4 h):Max. 5 %
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Informace o balení
25792_N-(1-NAPHTHYL)ETHYLENEDIAMINE DIHCL AR_BSE_REV0.pdf
Certifikáty BSE/TSE
spec_VWRC25792.120 spec_VWRC25792.120 spec_VWRC25792.130
Specifikace produktu
Frequently Bought Together