
N-(1-Naphthyl)ethylenediamine dihydrochloride 98+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1465-25-4
:
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
:259.18 g/mol
Merck Index:12,06496
Melting Pt:200 °C (dec.)
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige to pink Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.0 % (on dry substance)
- Water:≤5 % (K.F.)
- Loss on drying:≤5 % (1 g, 80°C, 4 h)
- Solubility:Passes test
- Sensitivity:(to sulfanilamide) Passes test
Specification Test Results
Related Information
Frequently Bought Together