



Malachite green oxalate for microscopy
3076.0025
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Identification:Passes test
:
:927.02 g/mol
:13,05722
:∼159 °C
:Ambient
:219-441-7
:2811
:6.1,III
:GHS08
:GHS07
:GHS05
:GHS09
:2437-29-8
:18015-76-4
:C₅₂H₅₄N₄O₁₂
:MFCD00011766