



Malachite green oxalate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2437-29-8
:18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766
:927.02 g/mol
Merck Index:14,5699
Melting Pt:164 °C (dec.)
Storage Temperature:Ambient
EINECS:219-441-7
UN:2811
ADR:6.1,III
:GHS05
:GHS08
:GHS07
:GHS09
Frequently Bought Together