

Diethylamine hydrochloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:660-68-4
:
Formula:C₄H₁₂ClN
MDL Number:MFCD00012499
:109.6 g/mol
Merck Index:13,03138
Boiling Pt:320 °C (1013 hPa)
Melting Pt:220…225 °C
Density:1.0475 g/cm³ (21 °C)
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:211-541-9
:GHS06
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:222 °C
- Molar Mass:109.60 g/mol
- Bulk Density:1100 kg/m3
- Boiling Point:320 - 330 °C
- pH:4.5 - 6.5 (10 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together