Dusičnan stříbrný concentrate for dilution to 1L, 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
Boiling Pt:100 °C
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,II
:GHS05
:GHS09
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together