Specifications
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:0501
- Environmentally Preferable:
Specifications
Specification Test Results
- Amount-of-substance concentration:0.01000 mol/l
Specification Test Results
About this item
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
:
:336.21 g/mol
:M-Clarity™ quality level = MQ100
EINECS:205-358-3
:GHS07
Storage Temperature:Ambient
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672