Order Entry
Czech Republic
ContactUsLinkComponent
 
L(+)-Histidine monohydrochloride monohydrate ≥98.5%, TECHNICAL
  24583.186
 :  
undefined
L(+)-Histidine monohydrochloride monohydrate ≥98.5%, TECHNICAL
  24583.186
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Assay:
    Min. 98.5 %

 

 

 :
 :209.63 g/mol
 :13,04741
 :259 °C
 :1.485 g/cm³ (20 °C)
 :Ambient
 :211-438-9
 :5934-29-2
 :C₆H₉N₃O₂·HCl·H₂O
 :MFCD00151027