Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate
EDQMC3290000
:
EDQM
:
L(+)-Cysteine hydrochloride monohydrate
EDQMC3290000
:
EDQM
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
175.64 g/mol
:
13,02810
:
168…170 °C
:
Ambient
:
200-157-7
:
7048-04-6
:
HSCH₂CH(NH₂)COOH·HCl·H₂O
:
MFCD00065606