Order Entry
Czech Republic
ContactUsLinkComponent
Iron(III) chloride 41% in aqueous solution, TECHNICAL
Corrosive
Irritant
Iron(III) chloride 41% in aqueous solution, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Iron(III) chloride 41% in aqueous solution, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    38 - 42 %

Specification Test Results

Frequently Bought Together