Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
Hydrazine hydrate 80% (∼51% w/w N₂H₄)
 
undefined
 
 
 
 
Hydrazine hydrate 80% (∼51% w/w N₂H₄)

 

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.4110 to 1.4140 (20°C, 589 nm)
  • Density:
    (20°C) 1.027 to 1.030 g/ml
  • Titration with HCl:
    50.5 to 51.6 % (N2H4)

 

 

Frequently Bought Together