Ammonia solution 10% (v/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Density:0.98
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
Related Information
Frequently Bought Together