Order Entry
Czech Republic
ContactUsLinkComponent
Hydroxylammonium chloride 10% in aqueous solution
Health
Hydroxylammonium chloride 10% in aqueous solution
Catalog # HACH81842
Supplier:  Hach
CAS Number:  
Health
Hydroxylammonium chloride 10% in aqueous solution
Catalog # HACH81842
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :69.49 g/mol
EINECS:226-798-2
 :GHS08
Storage Temperature:Ambient
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089