Tetramethylammonium hydroxide 25% in methanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Storage Temperature:Ambient
EINECS:200-882-9
UN:2924
ADR:3,II
Frequently Bought Together