



Tetramethylamoniumhydroxid 25% vodný roztok for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Density:1,02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-882-9
UN:1835
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS08
:GHS09
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:110 °C (1013 hPa)
- Vapor Pressure:23.33 hPa (20 °C)
- Density:1.02 g/cm3 (20 °C)
- pH:13 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Related Information
Frequently Bought Together