
Tetraethylammonium hydroxide 35% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-98-5
:
Formula:C₈H₂₁NO
MDL Number:MFCD00009024
:147.26 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:201-073-3
UN:3267
:GHS05
Frequently Bought Together