Order Entry
Czech Republic
ContactUsLinkComponent
Tetrabutylammonium hydroxide 40% (w/w) 1.5 M in water
Corrosive
Irritant
Tetrabutylammonium hydroxide 40% (w/w) 1.5 M in water
Certificates
undefined
Corrosive
Irritant
Tetrabutylammonium hydroxide 40% (w/w) 1.5 M in water
Certificates

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤150 APHA
  • Titration with HCl:
    38 to 42 %
  • Solubility:
    (5% in water) Clear colorless
  • Specific optical rotation:
    +20.0° to +24.0° (20°C, 589 nm) (c=1%, water)
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together