
Orthophosphoric acid ≥85%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS05
Frequently Bought Together