Order Entry
Czech Republic
ContactUsLinkComponent
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, J.T.Baker®
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, J.T.Baker®
Catalog # 7254.1000
Supplier:  Avantor
CAS Number:  
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, J.T.Baker®
Catalog # 7254.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302