Order Entry
Czech Republic
ContactUsLinkComponent
Silver nitrate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Irritant
Environment
Silver nitrate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4681.
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Environment
Silver nitrate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4681.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0998-0.1002
  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3082
ADR:9,III
 :GHS07
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414