Order Entry
Czech Republic
ContactUsLinkComponent
Silver nitrate 0.02 M AgNO3 (0.02N) ≥99% volumetric solution, Fluka™
Silver nitrate 0.02 M AgNO3 (0.02N) ≥99% volumetric solution, Fluka™
Catalog # 34296-1L
CAS Number:  
undefined
Silver nitrate 0.02 M AgNO3 (0.02N) ≥99% volumetric solution, Fluka™
Catalog # 34296-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :169.87 g/mol
Density:1.015 g/cm³
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414