


Phosphorus(III) bromide 1 mol/l in dichloromethane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-60-8
:
Formula:PBr₃
MDL Number:MFCD00011436
:270.69 g/mol
EINECS:232-178-2
UN:2922
:GHS07
:GHS08
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Titration Argentometric:0.9 to 1.1 M
Specification Test Results
Related Information
Frequently Bought Together