Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Hydrochloric acid 37%

Hydrochloric acid 37%

Plastic container, 20 L
Catalog # VWRS84539.440
Corrosive
Irritant
Supplier:   VWR Collection
Hydrochloric acid 37%
Catalog # VWRS84539.440
Supplier:  VWR Collection
CAS Number:  7647-01-0
Hydrochloric acid 37%

Plastic container, 20 L

CAS Number:  7647-01-0
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic container
  • Pk:
    20 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.