
Calcium chloride 1 mol/l in aqueous solution, Biotechnology Grade for biotechnology
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10035-04-8
:
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613
:147.01 g/mol
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identification:PASS
- Titration:0.95 - 1.05 M
Specification Test Results
Frequently Bought Together