Order Entry
Czech Republic
ContactUsLinkComponent
Calcium chloride 0.5 mol/l (1 N) in aqueous solution volumetric solution
Calcium chloride 0.5 mol/l (1 N) in aqueous solution volumetric solution
Catalog # REGECACL2051
Supplier:  REAGECON
CAS Number:  
Calcium chloride 0.5 mol/l (1 N) in aqueous solution volumetric solution
Catalog # REGECACL2051
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903