Order Entry
Czech Republic
ContactUsLinkComponent
Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31481.295
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31481.295
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413