Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 37%, EMPARTA® ACS analytical reagent
Corrosive
Irritant
Hydrochloric acid 37%, EMPARTA® ACS analytical reagent
Catalog # 1.01834.2011
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Hydrochloric acid 37%, EMPARTA® ACS analytical reagent
Catalog # 1.01834.2011
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -28 °C
  • Boiling Point:
    45 °C
  • Vapor Pressure:
    190 hPa (20 °C)
  • Density:
    1.19 g/cm3 (20 °C)
  • pH:
     - 1 (H2O, 20 °C)

Specification Test Results

About this item

 :
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
 :M-Clarity™ quality level = MQ200
 :36.46 g/mol
EINECS:231-595-7
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324