ContactUsLinkComponent
Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™
Corrosive
Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™
Certificates
Specification Test Results
  • Assay titr.:
    0.095 - 0.105 mol/l
  • Carbonate (CO₃):
    Max. 0.0015 mol/l
  • Traceable to NIST SRM:
    effective
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective