
Potassium hydroxide 0.1 M KOH (0.1N) volumetric solution, Fluka™
:1310-58-3
:
:KOH
:MFCD00003553
:56.11 g/mol
:Ambient
:215-181-3
:8,II
:GHS07
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together