Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Potassium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  7130.1000
 :  Avantor
 :  
 
Potassium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  7130.1000
 :  Avantor
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Titer (mol/l):
    0.0995 - 0.1005

 

 

 :
 :56.11 g/mol
 :Ambient
 :215-181-3
 :1814
 :8,II
 :GHS05
 :1310-58-3
 :KOH
 :MFCD00003553