


Methylamine 40% in aqueous solution, Sigma-Aldrich®
: Merck
:74-89-5
:
:CH₅N
:MFCD00008104
:31.06 g/mol
:3,II
:M-Clarity™ quality level = MQ200
:200-820-0
:1235
:GHS02
:GHS05
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals