Trimethylamine 31 - 35% (w/w) ∼4.2 mol/l in ethanol, Sigma-Aldrich®
: Merck
:75-50-3
:
:C₃H₉N
:MFCD00008327
:Ambient
:3,II
:59.11 g/mol
:200-875-0
:2924
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
: VWR Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Pipette Boxes, Square
: KOEHLER TECHNISCHE PRODUKTEN
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem