


Trimethylamine 33% (w/w) 4.2 M in ethanol, pure
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:75-50-3
:
Formula:C₃H₉N
MDL Number:MFCD00008327
Boiling Pt:50 °C
Density:0.75 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:200-875-0
UN:2924
ADR:3,II
:59.11 g/mol
:GHS07
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with HCl:32 to 34 %
Specification Test Results
Frequently Bought Together