
Glycolic acid ≥70% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-14-1
:
Formula:C₂H₄O₃
MDL Number:MFCD00004312
:76.05 g/mol
Density:1.262 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-180-5
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:10 °C
- Boiling Point:112 °C (1013 hPa)
- Vapor Pressure:27.5 hPa (25 °C)
- Density:1.26 g/cm3 (20 °C)
- pH:0.5 (H2O)
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Frequently Bought Together