Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
Health
Glycolic acid ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Catalog # 8.14186.1000
Supplier:  Merck
CAS Number:  
Corrosive
Health
Glycolic acid ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Catalog # 8.14186.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    10 °C
  • Boiling Point:
    112 °C (1013 hPa)
  • Density:
    1.262 g/cm3 (20 °C)
  • pH:
    0.1 (H2O, 25 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :76.05 g/mol
Density:1.262 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:201-180-5
 :GHS05
 :GHS08
CAS Number:79-14-1
Formula:C₂H₄O₃
MDL Number:MFCD00004312