ContactUsLinkComponent
Neutralising reagent
Neutralising reagent
Catalog # ENZOADI801475
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Neutralising reagent
Catalog # ENZOADI801475
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Neutralising reagent
  • Seq:
    02
Specifications
About this item
Neutralising buffer for low pH solutions and for use with cyclic nucleotide and isoprostane kits.