
N-Lauroylsarcosine sodium salt 20% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:137-16-6
:
Formula:H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number:MFCD00042728
:293.38 g/mol
:M-Clarity™ quality level = MQ200
EINECS:205-281-5
:GHS06
Storage Temperature:Ambient
Frequently Bought Together