Hydrochloric acid 0.5 M HCl (0.5N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:>100 °C (1.013 hPa)
Melting Pt:0 °C
Density:1.010 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS05
Specification Test Results
- Assay:0.4900 - 0.5100 N
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together