


1,5-Hexadiyne 50% in pentane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:628-16-0
:
Formula:C₆H₆
MDL Number:MFCD00014924
:78.11 g/mol
Density:0.805
Flash Pt:–2 °C (29 °F)
Storage Temperature:Ambient
EINECS:211-029-5
UN:3295
ADR:3,II
:Packaged under argon.
:GHS02
:GHS08
:GHS07
Related Information
Frequently Bought Together