Linoleic acid 60% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-33-3
:
Formula:C₁₈H₃₂O₂
MDL Number:MFCD00064241
:280.45 g/mol
Boiling Pt:229…230 °C (16 mmHg)
Melting Pt:–5 °C
Density:0.9 g/cm³
Flash Pt:>112 °C
Storage Temperature:Refrigerator
EINECS:200-470-9
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≤3 % C18 (sat.)
- Acid value:195 to 205 mg KOH/g
- Iodine value:130 to 148 g I2/100 g
Specification Test Results
Frequently Bought Together