Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl linoleate 65% (by GC), Technical Grade, Sigma-Aldrich®
Ethyl linoleate 65% (by GC), Technical Grade, Sigma-Aldrich®
Catalog # SIAL62262-500ML-F
Supplier:  Merck
CAS Number:  
Ethyl linoleate 65% (by GC), Technical Grade, Sigma-Aldrich®
Catalog # SIAL62262-500ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :308.5 g/mol
Storage Temperature:Freezer
EINECS:208-868-4
CAS Number:544-35-4
Formula:C₂₀H₃₆O₂
MDL Number:MFCD00009535