


DL-Limonene, Sigma-Aldrich®
Catalog # SIAL334111-4L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
About this item
:
:136.24 g/mol
Merck Index:12,05518
Boiling Pt:176 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.8462 g/cm³ (20 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:205-341-0
UN:2052
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:GHS09
CAS Number:138-86-3
:7705-14-8
Formula:C₁₀H₁₆
MDL Number:MFCD00062992