

(±)-3-Bromocyclohexene 95% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1521-51-3
:
Formula:C₆H₉Br
MDL Number:MFCD00013775
:161.04 g/mol
Boiling Pt:64…65 °C (15 mmHg)
Density:1.400
Flash Pt:54 °C (129 °F)
UN:1993
ADR:3,III
:Stabilised with propylene oxide
:GHS02
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together