1-Phenyl-2-propanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:698-87-3
:14898-87-4
Formula:C₆H₅CH₂CH(OH)CH₃
MDL Number:MFCD00004540
:136.19 g/mol
Boiling Pt:219 - 221 °C (1013 hPa)
Melting Pt:65 - 67 °C
Density:0.99 g/cm³
Flash Pt:85 °C
Storage Temperature:Ambient
EINECS:211-821-0
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5200 to 1.5220 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together