Order Entry
Czech Republic
ContactUsLinkComponent
 
(±)-3,7-Dimethyl-3-octanol, Sigma-Aldrich®
Catalog # SIALW306002-SAMPLE
Supplier:  Merck
CAS Number:  
(±)-3,7-Dimethyl-3-octanol, Sigma-Aldrich®
Catalog # SIALW306002-SAMPLE
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :158.28 g/mol
Boiling Pt:195.5…197 °C (1013 hPa)
Density:0.828 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
EINECS:201-133-9
CAS Number:78-69-3
Formula:C₁₀H₂₂O
MDL Number:MFCD00004482